missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, 98.0-100.5% TiO2, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 277370010

 View more versions of this product

Catalog No. AC277370010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

2900.0°C
1855.0°C
White
1 kg
TiO2
15, 9628
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
O=[Ti]=O
79.87
CHEBI:32234
98.0 to 100.5% TiO2
4.23
Titanium(IV) oxide
13463-67-7
Powder
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.88
Glass Bottle
4.2300g/mL

Chemical Identifiers

13463-67-7
79.87
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
CHEBI:32234
O=[Ti]=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 236-675-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only