missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, 98.0-100.5% TiO2, Thermo Scientific Chemicals

$128.33 - $838.90

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC277370010
SDS View Documents
Thermo Scientific Chemicals
277370010
1 kg Glass Bottle
Each for $128.33
 
AC277370100
SDS View Documents
Thermo Scientific Chemicals
277370100
10 kg Plastic drum
Each for $838.90
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

13463-67-7
79.87
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
CHEBI:32234
O=[Ti]=O

Specifications

2900.0°C
13463-67-7
Powder
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.88
Glass Bottle
4.2300g/mL
1855.0°C
White
1 kg
TiO2
15, 9628
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
O=[Ti]=O
79.87
CHEBI:32234
98.0 to 100.5% TiO2
4.23
Titanium(IV) oxide
Safety and Handling

Safety and Handling

EINECSNumber : 236-675-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only