missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 384290500

 View more versions of this product

Catalog No. AC384290500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

2900.0°C
1855.0°C
White to Yellow
Powder
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.88
1.5% max. (105°C, 2 hrs) typically
Plastic bottle
4.2300g/mL
Titanium(IV) oxide
13463-67-7
3.5 to 4.5 (4% dispersion)
50 g
TiO2
15, 9628
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
O=[Ti]=O
79.87
CHEBI:32234
≥99.5% (based on ignited material) (typically)
2.0% max. (1000°C, 2 hrs) (based on dried material) typically
4.23

Chemical Identifiers

13463-67-7
79.87
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

EINECSNumber : 236-675-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only