missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) oxide, Aeroxide™ P25
CAS: 13463-67-7 | O2Ti | 79.87 g/mol
$154.14 - $605.71
Chemical Identifiers
| CAS | 13463-67-7 |
|---|---|
| Molecular Formula | O2Ti |
| Molecular Weight (g/mol) | 79.87 |
| MDL Number | MFCD00011269 MFCD00210650 |
| InChI Key | GWEVSGVZZGPLCZ-UHFFFAOYSA-N |
| Synonym | titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance |
| PubChem CID | 26042 |
| ChEBI | CHEBI:32234 |
| IUPAC Name | dioxotitanium |
| SMILES | O=[Ti]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC384290500
|
Thermo Scientific Chemicals
384290500 |
50 g | Plastic bottle |
Each for $154.14
|
|
||||
|
AC384292500
|
Thermo Scientific Chemicals
384292500 |
250 g | Plastic bottle |
Each for $221.40
|
|
||||
|
AC384290010
|
Thermo Scientific Chemicals
384290010 |
1 kg | Plastic drum |
Each for $605.71
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
| 13463-67-7 | |
| 79.87 | |
| GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
| 26042 | |
| dioxotitanium |
| O2Ti | |
| MFCD00011269 MFCD00210650 | |
| titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
| CHEBI:32234 | |
| O=[Ti]=O |
Specifications
| 2900.0°C | |
| 13463-67-7 | |
| 3.5 to 4.5 (4% dispersion) | |
| 50 g | |
| TiO2 | |
| 15, 9628 | |
| Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf | |
| O=[Ti]=O | |
| 79.87 | |
| CHEBI:32234 | |
| ≥99.5% (based on ignited material) (typically) | |
| 2.0% max. (1000°C, 2 hrs) (based on dried material) typically | |
| 4.23 | |
| Titanium(IV) oxide |
| 1855.0°C | |
| White to Yellow | |
| Powder | |
| O2Ti | |
| MFCD00011269 MFCD00210650 | |
| titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
| GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
| dioxotitanium | |
| 26042 | |
| 79.88 | |
| 1.5% max. (105°C, 2 hrs) typically | |
| Plastic bottle | |
| 4.2300g/mL |
Safety and Handling
EINECSNumber : 236-675-5
RUO – Research Use Only