missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
T324-1
|
Thermo Fisher Scientific Formerly Fairlawn
T3241 |
1 L | Glass Bottle |
Each for $177.19
Case of 6 Each for $1,063.14
|
|
||||
T324-4
|
Thermo Fisher Scientific Formerly Fairlawn
T3244 |
4 L | Glass Bottle |
Each for $407.55
Case of 4 Each for $1,630.20
|
|
||||
T324-20
|
Thermo Fisher Scientific Formerly Fairlawn
BPT32420 |
20 L | Steel Pail |
Each for $1,330.00
|
|
||||
T324-200
|
Thermo Fisher Scientific Formerly Fairlawn
T324200 |
200 L | Steel Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
-95°C | |
Colorless | |
111°C | |
Pass Test | |
Liquid | |
≥99.5 % | |
MFCD00008512 | |
0.6 mPa/s at 20°C | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
≥99.5% |
108-88-3 | |
0.866g/cm³ | |
1 L | |
Glass Bottle | |
0.001% max. | |
C7H8 | |
29mbar at 20°C | |
0.03% max. | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
Toluene |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health: 2
Flammability: 3
Instability: 0
Signal Word: DANGER!
DOTInformation : DOT Class 3, : Flammable Liquid