missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Toluene (Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific Formerly Fairlawn T3244
Specifications
Toluene | |
108-88-3 | |
0.866g/cm³ | |
4 L | |
Glass Bottle | |
0.001% max. | |
C7H8 | |
29mbar at 20°C | |
0.03% max. | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 |
-95°C | |
Colorless | |
111°C | |
Pass Test | |
Liquid | |
≥99.5 % | |
MFCD00008512 | |
0.6 mPa/s at 20°C | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
≥99.5% |
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health: 2
Flammability: 3
Instability: 0
Signal Word: DANGER!
DOTInformation : DOT Class 3, : Flammable Liquid