missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediol, 97%

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol

Supplier:  Thermo Scientific Chemicals 161650100

 View more versions of this product

Catalog No. AC161650100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-1, 2-Cyclohexanediol
1460-57-7
White
134°C
98%
C6H12O2
MFCD00063611
17,91
Solubility in water: 375g/L (20 c)
O[C@@H]1CCCC[C@H]1O
116.16
8 mPa.s (110°C)
116.16
Crystalline Powder or Crystals
98%
100°C to 104°C
117°C (17.0 Mbar)
Authentic
Glass bottle
C6H10(OH)2
10 g
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PFURGBBHAOXLIO-PHDIDXHHSA-N
(1R,2R)-cyclohexane-1,2-diol
92886
CHEBI:16931
98%

Chemical Identifiers

1460-57-7
116.16
PFURGBBHAOXLIO-PHDIDXHHSA-N
92886
(1R,2R)-cyclohexane-1,2-diol
C6H12O2
MFCD00063611
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
CHEBI:16931
O[C@@H]1CCCC[C@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-956-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only