missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-1,2-Cyclohexanediol, 98%, Thermo Scientific Chemicals

$110.95 - $110.95

Chemical Identifiers

CAS 1460-57-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00063611
InChI Key PFURGBBHAOXLIO-PHDIDXHHSA-N
Synonym trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PubChem CID 92886
ChEBI CHEBI:16931
IUPAC Name (1R,2R)-cyclohexane-1,2-diol
SMILES O[C@@H]1CCCC[C@H]1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC161650100
SDS View Documents
Thermo Scientific Chemicals
161650100
10 g Glass bottle
Each for $110.95
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1460-57-7
116.16
PFURGBBHAOXLIO-PHDIDXHHSA-N
92886
(1R,2R)-cyclohexane-1,2-diol
C6H12O2
MFCD00063611
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
CHEBI:16931
O[C@@H]1CCCC[C@H]1O

Specifications

1460-57-7
White
134°C
98%
C6H12O2
MFCD00063611
17,91
Solubility in water: 375g/L (20 c)
O[C@@H]1CCCC[C@H]1O
116.16
8 mPa.s (110°C)
116.16
Crystalline Powder or Crystals
100°C to 104°C
117°C (17.0 Mbar)
Authentic
Glass bottle
C6H10(OH)2
10 g
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PFURGBBHAOXLIO-PHDIDXHHSA-N
(1R,2R)-cyclohexane-1,2-diol
92886
CHEBI:16931
98%
trans-1, 2-Cyclohexanediol, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-956-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only