missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octene, 97%

CAS: 13389-42-9 | C8H16 | 112.22 g/mol

$158.00 - $511.95

Chemical Identifiers

CAS 13389-42-9
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00009532
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
Synonym trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
PubChem CID 5364448
IUPAC Name (E)-oct-2-ene
SMILES CCCCC\C=C\C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1260606
View Documents
Thermo Scientific Chemicals
L1260606
5 g
Each for $158.00
Only null left
 
AAL1260614
View Documents
Thermo Scientific Chemicals
L1260614
25 g
Each for $511.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13389-42-9
112.22
ILPBINAXDRFYPL-HWKANZROSA-N
5364448
CCCCC\C=C\C
C8H16
MFCD00009532
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
(E)-oct-2-ene

Specifications

13389-42-9
0.718
14°C (57°F)
1.413
5 g
1719497
ILPBINAXDRFYPL-HWKANZROSA-N
(E)-oct-2-ene
5364448
97%
-89°C to -86°C
122°C to 123°C
C8H16
MFCD00009532
UN3295
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
CCCCC\C=C\C
112.22
112.22
trans-2-Octene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 236-463-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only