Learn More
trans-2-Pentene, 99%, Thermo Scientific Chemicals
$233.55 - $233.55
Chemical Identifiers
CAS | 646-04-8 |
---|---|
Molecular Formula | C5H10 |
Molecular Weight (g/mol) | 70.135 |
MDL Number | MFCD00009384 |
InChI Key | QMMOXUPEWRXHJS-HWKANZROSA-N |
Synonym | trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e |
PubChem CID | 5326161 |
IUPAC Name | (E)-pent-2-ene |
SMILES | CCC=CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0942106
|
Thermo Scientific Chemicals
L0942106 |
5 g |
Each for $233.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
646-04-8 | |
70.135 | |
QMMOXUPEWRXHJS-HWKANZROSA-N | |
5326161 | |
CCC=CC |
C5H10 | |
MFCD00009384 | |
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e | |
(E)-pent-2-ene |
Specifications
646-04-8 | |
0.649 | |
−45°C (−49°F) | |
C5H10 | |
MFCD00009384 | |
UN3295 | |
14,7123 | |
QMMOXUPEWRXHJS-HWKANZROSA-N | |
(E)-pent-2-ene | |
5326161 | |
99% |
-136°C | |
36°C to 37°C | |
Petroleum-like | |
1.379 | |
5 g | |
1718795 | |
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e | |
CCC=CC | |
70.135 | |
70.14 | |
trans-2-Pentene |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 211-461-4
RTECSNumber : SB2184500
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only