Learn More
trans-4-Methylcyclohexylamine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 455920050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
trans-4-Methylcyclohexylamine | |
0.8500g/mL | |
Authentic | |
C7H15N | |
1.4500 to 1.5480 | |
0.85 | |
KSMVBYPXNKCPAJ-UHFFFAOYSA-N | |
4-methylcyclohexan-1-amine | |
80604 | |
99% |
2523-55-9 | |
151.0°C to 154.0°C | |
98.5% min. (GC) | |
CH3C6H10NH2 | |
5g | |
4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine | |
CC1CCC(CC1)N | |
113.2 | |
113.2 |
Chemical Identifiers
2523-55-9 | |
113.2 | |
4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine | |
4-methylcyclohexan-1-amine |
C7H15N | |
KSMVBYPXNKCPAJ-UHFFFAOYSA-N | |
80604 | |
CC1CCC(CC1)N |
Safety and Handling
GHS H Statement Flammable liquid and vapour. Causes severe skin burns and eye damage.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
RUO â Research Use Only