missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans,trans-2,4-Hexadien-1-ol, 99%, stabilized, Thermo Scientific Chemicals
$122.44 - $407.33
Chemical Identifiers
CAS | 17102-64-6 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.15 |
MDL Number | MFCD00002925 |
InChI Key | MEIRRNXMZYDVDW-MQQKCMAXSA-N |
Synonym | sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol |
PubChem CID | 641256 |
IUPAC Name | (2E,4E)-hexa-2,4-dien-1-ol |
SMILES | C\C=C\C=C\CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC325570100
|
Thermo Scientific Chemicals
325570100 |
10 g |
Each for $122.44
|
|
|||||
AC325570500
|
Thermo Scientific Chemicals
325570500 |
50 g |
Each for $407.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17102-64-6 | |
98.15 | |
MEIRRNXMZYDVDW-MQQKCMAXSA-N | |
641256 | |
C\C=C\C=C\CO |
C6H10O | |
MFCD00002925 | |
sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol | |
(2E,4E)-hexa-2,4-dien-1-ol |
Specifications
17102-64-6 | |
Colorless to Yellow | |
80.0°C (12.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4990 to 1.5010 | |
MFCD00002925 | |
0.87 | |
sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol | |
MEIRRNXMZYDVDW-MQQKCMAXSA-N | |
(2E,4E)-hexa-2,4-dien-1-ol | |
0.1% alpha-tocopherol | |
98.14 | |
Liquid After Melting |
30.5°C to 31.5°C | |
0.8700g/mL | |
72°C | |
98.5% min. (GC) | |
C6H10O | |
CH3CH=CHCH=CHCH2OH | |
10 g | |
15,8848 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and oils | |
C\C=C\C=C\CO | |
98.15 | |
641256 | |
99% | |
trans, trans-2, 4-Hexadien-1-ol |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 241-173-4
RUO – Research Use Only