Learn More
Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A12649AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri-n-butylphosphine is used as a catalyst modifier in the cobalt-catalyzed hydroformylation of alkenes for industrial use. It is also used as a strong reducing agent and polymerization cross-linking catalyst. It is a used as an intermediate for phase transfer catalysts.
Solubility
Soluble in organic solvent such as heptane. Not miscible with water.
Notes
Air and moisture Sensitive. Store away from water/moisture and air. Store in cool dry place in tightly closed container.
Specifications
Tri-n-butylphosphine | |
-65°C | |
240°C | |
Unpleasant | |
1.462 | |
100 mL | |
1738261 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
tributylphosphane | |
13831 | |
95% |
998-40-3 | |
0.82 | |
>93°C (199°F) | |
C12H27P | |
MFCD00009462 | |
UN3254 | |
Air and moisture sensitive | |
Soluble in organic solvent such as heptane. Not miscible with water. | |
CCCCP(CCCC)CCCC | |
202.32 | |
202.32 |
Chemical Identifiers
998-40-3 | |
202.32 | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
13831 | |
CCCCP(CCCC)CCCC |
C12H27P | |
MFCD00009462 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
tributylphosphane |
Safety and Handling
GHS H Statement
H250-H252-H314-H318-H302-H312
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P210-P222-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H250-H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: TRIBUTYLPHOSPHANE
EINECSNumber : 213-651-2
RTECSNumber : SZ3270000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only