Learn More
Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals
$68.70 - $591.85
Chemical Identifiers
CAS | 998-40-3 |
---|---|
Molecular Formula | C12H27P |
Molecular Weight (g/mol) | 202.32 |
MDL Number | MFCD00009462 |
InChI Key | TUQOTMZNTHZOKS-UHFFFAOYSA-N |
Synonym | tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp |
PubChem CID | 13831 |
IUPAC Name | tributylphosphane |
SMILES | CCCCP(CCCC)CCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA12649AC
|
Thermo Scientific Chemicals
A12649AC |
25 mL |
Each for $68.70
|
|
|||||
AAA12649AE
|
Thermo Scientific Chemicals
A12649AE |
100 mL |
Each for $181.36
|
|
|||||
AAA12649AP
|
Thermo Scientific Chemicals
A12649AP |
500 mL |
Each for $591.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri-n-butylphosphine is used as a catalyst modifier in the cobalt-catalyzed hydroformylation of alkenes for industrial use. It is also used as a strong reducing agent and polymerization cross-linking catalyst. It is a used as an intermediate for phase transfer catalysts.
Solubility
Soluble in organic solvent such as heptane. Not miscible with water.
Notes
Air and moisture Sensitive. Store away from water/moisture and air. Store in cool dry place in tightly closed container.
Chemical Identifiers
998-40-3 | |
202.32 | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
13831 | |
CCCCP(CCCC)CCCC |
C12H27P | |
MFCD00009462 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
tributylphosphane |
Specifications
998-40-3 | |
0.82 | |
>93°C (199°F) | |
C12H27P | |
MFCD00009462 | |
UN3254 | |
Air and moisture sensitive | |
Soluble in organic solvent such as heptane. Not miscible with water. | |
CCCCP(CCCC)CCCC | |
202.32 | |
202.32 | |
Tri-n-butylphosphine |
-65°C | |
240°C | |
Unpleasant | |
1.462 | |
25 mL | |
1738261 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
tributylphosphane | |
13831 | |
95% |
Safety and Handling
GHS H Statement
H250-H252-H314-H318-H302-H312
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P210-P222-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H250-H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: TRIBUTYLPHOSPHANE
EINECSNumber : 213-651-2
RTECSNumber : SZ3270000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only