missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triacetin, 99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210309890

 View more versions of this product

Catalog No. ICN10309890


Explore available promotions

Description

Description

Specifications

Specifications

Triacetin
Colorless
99%
0.5 L
URAYPUMNDPQOKB-UHFFFAOYSA-N
2,3-diacetyloxypropyl acetate
5541
218.2
102-76-1
1.159g/cm3 at 20°C
C9H14O6
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
CC(=O)OCC(COC(=O)C)OC(=O)C
218.205
CHEBI:9661
Semi-viscous Liquid

Chemical Identifiers

102-76-1
218.205
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
CHEBI:9661
CC(=O)OCC(COC(=O)C)OC(=O)C
C9H14O6
URAYPUMNDPQOKB-UHFFFAOYSA-N
5541
2,3-diacetyloxypropyl acetate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-051-9

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions