missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triacetin, 99%, MP Biomedicals™
$195.62 - $195.62
Chemical Identifiers
CAS | 102-76-1 |
---|---|
Molecular Formula | C9H14O6 |
Molecular Weight (g/mol) | 218.205 |
InChI Key | URAYPUMNDPQOKB-UHFFFAOYSA-N |
Synonym | triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine |
PubChem CID | 5541 |
ChEBI | CHEBI:9661 |
IUPAC Name | 2,3-diacetyloxypropyl acetate |
SMILES | CC(=O)OCC(COC(=O)C)OC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10309890
|
MP Biomedicals Inc
0210309890 |
0.5 L |
Each for $195.62
|
|
|||||
Chemical Identifiers
102-76-1 | |
218.205 | |
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine | |
CHEBI:9661 | |
CC(=O)OCC(COC(=O)C)OC(=O)C |
C9H14O6 | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
5541 | |
2,3-diacetyloxypropyl acetate |
Specifications
102-76-1 | |
1.159g/cm3 at 20°C | |
C9H14O6 | |
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine | |
CC(=O)OCC(COC(=O)C)OC(=O)C | |
218.205 | |
CHEBI:9661 | |
Semi-viscous Liquid |
Colorless | |
99% | |
0.5 L | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
2,3-diacetyloxypropyl acetate | |
5541 | |
218.2 | |
Triacetin |
Safety and Handling
EINECSNumber : 203-051-9
Recommended Storage : Store at room temperature.