missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tributyrin, 97%, Thermo Scientific Chemicals

Tributyrin, 97%, C15H26O6, CAS Number-60-01-5, butyrin, butyryl triglyceride, glycerol tributanoate, glycerol tributyrate, glyceroltributyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, tributin, tributyrin, tri-n-butyrin, 100mL, Liquid, 96% min. (GC), 02, 273, 305.0 deg.C to 310.0 deg.C

Supplier:  Thermo Scientific Chemicals 150881000

 View more versions of this product

Catalog No. AC150881000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tributyrin
60-01-5
Clear colorless to light yellow
305.0°C to 310.0°C
Conforms
Glass bottle
1.4340 to 1.437
MFCD00009392
02, 273
15, 9785
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC
302.36
CHEBI:35020
97%
98%
-75.0°C
1.0335g/mL
173°C
96% min. (GC)
C15H26O6
(CH3CH2CH2CO2CH2)2CH(O2CCH2CH2CH3)
100 mL
1.0335
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
UYXTWWCETRIEDR-UHFFFAOYSA-N
2,3-di(butanoyloxy)propyl butanoate
6050
302.36
Liquid

Chemical Identifiers

60-01-5
302.36
UYXTWWCETRIEDR-UHFFFAOYSA-N
6050
2,3-di(butanoyloxy)propyl butanoate
C15H26O6
MFCD00009392
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
CHEBI:35020
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-451-5

RTECSNumber : ET7350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only