missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tributyrin, 97%, Thermo Scientific Chemicals
Tributyrin, 97%, C15H26O6, CAS Number-60-01-5, butyrin, butyryl triglyceride, glycerol tributanoate, glycerol tributyrate, glyceroltributyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, tributin, tributyrin, tri-n-butyrin, 100mL, Liquid, 96% min. (GC), 02, 273, 305.0 deg.C to 310.0 deg.C
$97.75 - $237.56
Chemical Identifiers
CAS | 60-01-5 |
---|---|
Molecular Formula | C15H26O6 |
Molecular Weight (g/mol) | 302.36 |
MDL Number | MFCD00009392 |
InChI Key | UYXTWWCETRIEDR-UHFFFAOYSA-N |
Synonym | tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin |
PubChem CID | 6050 |
ChEBI | CHEBI:35020 |
IUPAC Name | 2,3-di(butanoyloxy)propyl butanoate |
SMILES | CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC150881000
|
Thermo Scientific Chemicals
150881000 |
100 mL | Glass bottle |
N/A
|
|
||||
AC150882500
|
Thermo Scientific Chemicals
150882500 |
250 mL | Glass bottle |
N/A
|
|
||||
AC150880010
|
Thermo Scientific Chemicals
150880010 |
1 L | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-01-5 | |
302.36 | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
6050 | |
2,3-di(butanoyloxy)propyl butanoate |
C15H26O6 | |
MFCD00009392 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
CHEBI:35020 | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Specifications
60-01-5 | |
Clear colorless to light yellow | |
305.0°C to 310.0°C | |
Conforms | |
Glass bottle | |
1.4340 to 1.437 | |
MFCD00009392 | |
02, 273 | |
15, 9785 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
302.36 | |
CHEBI:35020 | |
97% | |
Tributyrin, 98% |
-75.0°C | |
1.0335g/mL | |
173°C | |
96% min. (GC) | |
C15H26O6 | |
(CH3CH2CH2CO2CH2)2CH(O2CCH2CH2CH3) | |
100 mL | |
1.0335 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
2,3-di(butanoyloxy)propyl butanoate | |
6050 | |
302.36 | |
Liquid |
Safety and Handling
EINECSNumber : 200-451-5
RTECSNumber : ET7350000
TSCA : TSCA
RUO – Research Use Only