Learn More
Tributyrin, 97%, Thermo Scientific Chemicals
Tributyrin, 97%, C15H26O6, CAS Number-60-01-5, butyrin, butyryl triglyceride, glycerol tributanoate, glycerol tributyrate, glyceroltributyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, tributin, tributyrin, tri-n-butyrin, 100mL, Liquid, 96% min. (GC), 02, 273, 305.0 deg.C to 310.0 deg.C
Supplier: Thermo Scientific Chemicals 150882500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tributyrin | |
60-01-5 | |
Clear colorless to light yellow | |
305°C to 310°C | |
Conforms | |
Glass bottle | |
1.4340 to 1.437 | |
MFCD00009392 | |
02, 273 | |
15, 9785 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
302.36 | |
CHEBI:35020 | |
97% |
98% | |
-75°C | |
1.0335g/mL | |
173°C | |
96% min. (GC) | |
C15H26O6 | |
(CH3CH2CH2CO2CH2)2CH(O2CCH2CH2CH3) | |
250 mL | |
1.0335 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
2,3-di(butanoyloxy)propyl butanoate | |
6050 | |
302.36 | |
Liquid |
Chemical Identifiers
60-01-5 | |
302.36 | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
6050 | |
2,3-di(butanoyloxy)propyl butanoate |
C15H26O6 | |
MFCD00009392 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
CHEBI:35020 | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Safety and Handling
EINECSNumber : 200-451-5
RTECSNumber : ET7350000
TSCA : TSCA
RUO – Research Use Only