Learn More
Trichloroacetic Acid (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLA3223
Specifications
Trichloroacetic Acid | |
52°C | |
196°C | |
≥99.0 % | |
CCl3COOH | |
1839 | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
2,2,2-trichloroacetic acid | |
6421 | |
163.39 | |
Certified ACS | |
3 kg | |
Glass Bottle | |
1.2mbar at 50°C, 0.08mbar at 25°C |
1.2 | |
76-03-9 | |
White | |
C2HCl3O2 | |
MFCD00004177 | |
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
C(=O)(C(Cl)(Cl)Cl)O | |
163.378 | |
CHEBI:30956 | |
≥99.0% | |
1.620g/cm³ | |
0.03% max. | |
Solid |
Chemical Identifiers
76-03-9 | |
163.378 | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
6421 | |
2,2,2-trichloroacetic acid |
C2HCl3O2 | |
MFCD00004177 | |
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
CHEBI:30956 | |
C(=O)(C(Cl)(Cl)Cl)O |
Safety and Handling
DANGER!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:3
Flammability:1
Instability:1
DOTInformation : DOT Class 8, : Corrosive