missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichloroacetic Acid (Crystalline/Certified ACS), Fisher Chemical™

$371.33 - $2680.25

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A322-100
SDS View Documents Promotion Details
Thermo Fisher Scientific
FLA322100
100 g Glass Bottle
Each for $371.33
Case of 6 Each for $2,227.98
 
A322-500
SDS View Documents Promotion Details
Thermo Fisher Scientific
FLA322500
500 g Glass Bottle
Each for $645.42
Case of 6 Each for $3,872.52
 
A322-3
SDS View Documents
Thermo Fisher Scientific
FLA3223
3 kg Glass Bottle
Each for $2,680.25
Case of 4 Each for $10,721.00
 
Description

Description

Specifications

Chemical Identifiers

76-03-9
163.378
YNJBWRMUSHSURL-UHFFFAOYSA-N
6421
2,2,2-trichloroacetic acid
C2HCl3O2
MFCD00004177
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
CHEBI:30956
C(=O)(C(Cl)(Cl)Cl)O

Specifications

1.2
76-03-9
White
C2HCl3O2
MFCD00004177
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
C(=O)(C(Cl)(Cl)Cl)O
163.378
CHEBI:30956
≥99.0%
1.620g/cm³
0.03% max.
Solid
Trichloroacetic Acid
52°C
196°C
≥99.0 %
CCl3COOH
1839
YNJBWRMUSHSURL-UHFFFAOYSA-N
2,2,2-trichloroacetic acid
6421
163.39
Certified ACS
100 g
Glass Bottle
1.2mbar at 50°C, 0.08mbar at 25°C
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:1
Instability:1

DOTInformation : DOT Class 8, : Corrosive

SDS
missing translation for 'documents'

missing translation for 'documents'