missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl 4-phosphonocrotonate, cis + trans, 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1099514

 View more versions of this product

Catalog No. AAA1099514


Explore available promotions

Description

Description

Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Triethyl 4-phosphonocrotonate
10236-14-3
133°C to 135°C (0.4 mmHg)
C10H19O5P
MFCD00009192
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
CCOC(=O)\C=C\CP(=O)(OCC)OCC
250.23
250.23
cis + trans
1.13
>110°C (230°F)
1.455
25 g
LXLODBXSCRTXFG-BQYQJAHWSA-N
ethyl (E)-4-diethoxyphosphorylbut-2-enoate
5365783
94%

Chemical Identifiers

10236-14-3
250.23
LXLODBXSCRTXFG-BQYQJAHWSA-N
5365783
CCOC(=O)\C=C\CP(=O)(OCC)OCC
C10H19O5P
MFCD00009192
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
ethyl (E)-4-diethoxyphosphorylbut-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only