missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl 4-phosphonocrotonate, cis + trans, 94%, Thermo Scientific Chemicals

$199.05 - $372.53

Chemical Identifiers

CAS 10236-14-3
Molecular Formula C10H19O5P
Molecular Weight (g/mol) 250.23
MDL Number MFCD00009192
InChI Key LXLODBXSCRTXFG-BQYQJAHWSA-N
Synonym triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
PubChem CID 5365783
IUPAC Name ethyl (E)-4-diethoxyphosphorylbut-2-enoate
SMILES CCOC(=O)\C=C\CP(=O)(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1099509
View Documents
Thermo Scientific Chemicals
A1099509
10 g
Each for $199.05
 
AAA1099514
View Documents
Thermo Scientific Chemicals
A1099514
25 g
Each for $372.53
 
Description

Description

Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

10236-14-3
250.23
LXLODBXSCRTXFG-BQYQJAHWSA-N
5365783
CCOC(=O)\C=C\CP(=O)(OCC)OCC
C10H19O5P
MFCD00009192
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
ethyl (E)-4-diethoxyphosphorylbut-2-enoate

Specifications

10236-14-3
133°C to 135°C (0.4 mmHg)
C10H19O5P
MFCD00009192
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
CCOC(=O)\C=C\CP(=O)(OCC)OCC
250.23
250.23
Triethyl 4-phosphonocrotonate, cis + trans
1.13
>110°C (230°F)
1.455
10 g
LXLODBXSCRTXFG-BQYQJAHWSA-N
ethyl (E)-4-diethoxyphosphorylbut-2-enoate
5365783
94%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only