missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethyl Citrate 99.0+%, TCI America™
$28.21 - $99.54
Chemical Identifiers
CAS | 77-93-0 |
---|---|
Molecular Formula | C12H20O7 |
Molecular Weight (g/mol) | 276.285 |
MDL Number | MFCD00009201 |
InChI Key | DOOTYTYQINUNNV-UHFFFAOYSA-N |
Synonym | triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester |
PubChem CID | 6506 |
IUPAC Name | triethyl 2-hydroxypropane-1,2,3-tricarboxylate |
SMILES | CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O |
Chemical Identifiers
77-93-0 | |
276.285 | |
DOOTYTYQINUNNV-UHFFFAOYSA-N | |
6506 | |
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O |
C12H20O7 | |
MFCD00009201 | |
triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester | |
triethyl 2-hydroxypropane-1,2,3-tricarboxylate |
Specifications
77-93-0 | |
Yellow | |
C12H20O7 | |
25 g | |
DOOTYTYQINUNNV-UHFFFAOYSA-N | |
triethyl 2-hydroxypropane-1,2,3-tricarboxylate | |
6506 | |
≥99.0% (GC) | |
Triethyl Citrate |
-45°C | |
294°C | |
MFCD00009201 | |
triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester | |
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O | |
276.285 | |
276.29 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-1320
RTECSNumber : GE8050000
TSCA : Yes