missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl orthoformate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A13587AP

 View more versions of this product

Catalog No. AAA13587AP

Explore available promotions

Description

Description

Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

Solubility
Miscible with alcohols, acetone, ethers and chloroform. Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with acids and strong oxidizing agents.
Specifications

Specifications

Triethyl Orthoformate
°C to 61°C
144°C to 146°C
98%
1.391
MFCD00009230
UN2524
Moisture sensitive
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
GKASDNZWUGIAMG-UHFFFAOYSA-N
diethoxymethoxyethane
31214
98%
122-51-0
0.891
30°C (86°F)
C7H16O3
HC(OCH2CH3)3
500 mL
605384
14,6884
Miscible with alcohols,acetone,ethers and chloroform. Slightly miscible with water.
CCOC(OCC)OCC
148.202
148.2
Liquid

Chemical Identifiers

122-51-0
148.202
GKASDNZWUGIAMG-UHFFFAOYSA-N
31214
CCOC(OCC)OCC
C7H16O3
MFCD00009230
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
diethoxymethoxyethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYL ORTHOFORMATE

EINECSNumber : 204-550-4

RTECSNumber : RM6475000

TSCA : Yes

storageNote1 : Moisture sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only