missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, 98%

CAS: 122-51-0 | C7H16O3 | 148.202 g/mol

$98.74 - $907.60

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
MDL Number MFCD00009230
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA13587AP
View Documents
Thermo Scientific Chemicals
A13587AP
500 mL
Each for $98.74
Only null left
 
AAA135870F
View Documents
Thermo Scientific Chemicals
A135870F
2500 mL
Each for $296.03
Only null left
 
AAA135870D
View Documents
Thermo Scientific Chemicals
A135870D
10,000 mL
Each for $907.60
Only null left
 
Description

Description

Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

Solubility
Miscible with alcohols, acetone, ethers and chloroform. Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with acids and strong oxidizing agents.
Specifications

Chemical Identifiers

122-51-0
148.202
GKASDNZWUGIAMG-UHFFFAOYSA-N
31214
CCOC(OCC)OCC
C7H16O3
MFCD00009230
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
diethoxymethoxyethane

Specifications

122-51-0
0.891
30°C (86°F)
C7H16O3
HC(OCH2CH3)3
500 mL
605384
14,6884
Miscible with alcohols,acetone,ethers and chloroform. Slightly miscible with water.
CCOC(OCC)OCC
148.202
148.2
Liquid
°C to 61°C
144°C to 146°C
98%
1.391
MFCD00009230
UN2524
Moisture sensitive
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
GKASDNZWUGIAMG-UHFFFAOYSA-N
diethoxymethoxyethane
31214
98%
Triethyl Orthoformate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYL ORTHOFORMATE

EINECSNumber : 204-550-4

RTECSNumber : RM6475000

TSCA : Yes

storageNote1 : Moisture sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only