missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylamine 99.0+%, TCI America™

$31.14 - $50.17

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T042425ML
View Documents
TCI America
T042425ML
25 mL
Each for $31.14
 
T0424100ML
View Documents
TCI America
T0424100ML
100 mL
Each for $32.87
 
T0424500ML
View Documents
TCI America
T0424500ML
500 mL
Each for $50.17
 
Description

Description

Specifications

Chemical Identifiers

121-44-8
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine
C6H15N
MFCD00009051
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC

Specifications

121-44-8
Colorless
C6H15N
25 mL
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CCN(CC)CC
101.193
CHEBI:35026
≥99.0% (GC,T)
Triethylamine
-115°C
89°C
MFCD00009051
1296
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
101.19
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0141

RTECSNumber : YE0175000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'