missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethylamine 99.0+%, TCI America™
Supplier: TCI America T0424100ML
Specifications
Triethylamine | |
-115°C | |
89°C | |
MFCD00009051 | |
1296 | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
N,N-diethylethanamine | |
8471 | |
101.19 | |
Liquid |
121-44-8 | |
Colorless | |
C6H15N | |
100 mL | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
CCN(CC)CC | |
101.193 | |
CHEBI:35026 | |
≥99.0% (GC,T) |
Chemical Identifiers
121-44-8 | |
101.193 | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
8471 | |
N,N-diethylethanamine |
C6H15N | |
MFCD00009051 | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
CHEBI:35026 | |
CCN(CC)CC |
Safety and Handling
EINECSNumber : (2)-0141
RTECSNumber : YE0175000
TSCA : Yes