Learn More
Triethylamine trihydrofluoride, ca. 37% HF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 350621000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Triethylamine trihydrofluoride | |
-29.0°C | |
0.9890g/mL | |
87°C | |
97% | |
C6H15N·3HF | |
(C2H5)3N·3HF | |
100 mL | |
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3 | |
IKGLACJFEHSFNN-UHFFFAOYSA-N | |
N,N-diethylethanamine;trihydrofluoride | |
175505 | |
97% |
73602-61-6 | |
Yellow to Brown | |
70.0°C (15.0 mmHg) | |
Authentic | |
Plastic bottle | |
1.3860 to 1.3920 | |
MFCD00043294 | |
0.989 | |
Solubility in water: soluble | |
CCN(CC)CC.F.F.F | |
161.21 | |
161.21 | |
Liquid |
Chemical Identifiers
73602-61-6 | |
161.21 | |
IKGLACJFEHSFNN-UHFFFAOYSA-N | |
175505 | |
CCN(CC)CC.F.F.F |
C6H15N·3HF | |
MFCD00043294 | |
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3 | |
N,N-diethylethanamine;trihydrofluoride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 277-550-5
RUO – Research Use Only