missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylamine trihydrofluoride, ca. 37% HF, Thermo Scientific Chemicals

$132.50 - $347.26

Chemical Identifiers

CAS 73602-61-6
Molecular Formula C6H153HF
Molecular Weight (g/mol) 161.21
MDL Number MFCD00043294
InChI Key IKGLACJFEHSFNN-UHFFFAOYSA-N
Synonym triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3
PubChem CID 175505
IUPAC Name N,N-diethylethanamine;trihydrofluoride
SMILES CCN(CC)CC.F.F.F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350620250
SDS View Documents
Thermo Scientific Chemicals
350620250
25 mL Plastic bottle
Each for $132.50
 
AC350621000
SDS View Documents
Thermo Scientific Chemicals
350621000
100 mL Plastic bottle
Each for $347.26
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

73602-61-6
161.21
IKGLACJFEHSFNN-UHFFFAOYSA-N
175505
CCN(CC)CC.F.F.F
C6H153HF
MFCD00043294
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3
N,N-diethylethanamine;trihydrofluoride

Specifications

73602-61-6
Yellow to Brown
70.0°C (15.0 mmHg)
Authentic
Plastic bottle
1.3860 to 1.3920
MFCD00043294
0.989
Solubility in water: soluble
CCN(CC)CC.F.F.F
161.21
161.21
Liquid
-29.0°C
0.9890g/mL
87°C
97%
C6H153HF
(C2H5)3N·3HF
25 mL
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3
IKGLACJFEHSFNN-UHFFFAOYSA-N
N,N-diethylethanamine;trihydrofluoride
175505
97%
Triethylamine trihydrofluoride, ca. 37% HF
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 277-550-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only