missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylenetetramine, 60%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 344070500

 View more versions of this product

Catalog No. AC344070500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Triethylenetetramine
112-24-3
0.9800g/mL
129°C
55% min. (GC)
C6H18N4
H2NCH2CH2NHCH2CH2NHCH2CH2NH2
50 g
01,1204
15,9828
Solubility in water: soluble in water. Other solubilities: soluble in alcohol
C(CNCCNCCN)N
146.24
CHEBI:39501
60%
Liquid
60%
-35.0°C
280.0°C
Authentic
Glass bottle
1.4960 to 1.5000
MFCD00008169
04,255
0.98
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
VILCJCGEZXAXTO-UHFFFAOYSA-N
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
5565
146.24
Technical

Chemical Identifiers

112-24-3
146.24
VILCJCGEZXAXTO-UHFFFAOYSA-N
5565
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
C6H18N4
MFCD00008169
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
CHEBI:39501
C(CNCCNCCN)N
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful in contact with skin.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 203-950-6

RTECSNumber : YE6650000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only