missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylenetetramine, 60%, Thermo Scientific Chemicals

$48.57 - $80.96

Chemical Identifiers

CAS 112-24-3
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.24
MDL Number MFCD00008169
InChI Key VILCJCGEZXAXTO-UHFFFAOYSA-N
Synonym triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
PubChem CID 5565
ChEBI CHEBI:39501
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
SMILES C(CNCCNCCN)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC344070500
SDS View Documents
Thermo Scientific Chemicals
344070500
50 g
Each for $48.57
 
AC344075000
SDS View Documents
Thermo Scientific Chemicals
344075000
500 g
Each for $80.96
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-24-3
146.24
VILCJCGEZXAXTO-UHFFFAOYSA-N
5565
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
C6H18N4
MFCD00008169
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
CHEBI:39501
C(CNCCNCCN)N

Specifications

112-24-3
0.9800g/mL
129°C
55% min. (GC)
C6H18N4
H2NCH2CH2NHCH2CH2NHCH2CH2NH2
50 g
01,1204
15,9828
Solubility in water: soluble in water. Other solubilities: soluble in alcohol
C(CNCCNCCN)N
146.24
CHEBI:39501
60%
Liquid
-35.0°C
280.0°C
Authentic
Glass bottle
1.4960 to 1.5000
MFCD00008169
04,255
0.98
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
VILCJCGEZXAXTO-UHFFFAOYSA-N
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
5565
146.24
Technical
Triethylenetetramine, 60%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful in contact with skin.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 203-950-6

RTECSNumber : YE6650000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only