missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Acros Organics, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0

Supplier:  Thermo Scientific Chemicals 147812501

 View more versions of this product

Catalog No. AC147812501


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
Specifications

Specifications

Trifluoroacetic anhydride
-63.5°C
39.1°C
99% min. (GC)
C4F6O3
(CF3CO)2O
250 g
01,1221; 03,308; 05,701; 06,616; 07,389; 09,484; 12,530; 14,151; 15,14; 01,373
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
9845
141mbar at 20°C
Liquid
407-25-0
1.5100g/mL
Authentic
Glass bottle
1.3
MFCD00000416
02, II, 186
1.51
Solubility in water: hydrolysis. Other solubilities: miscible with polar organic solvents, partically soluble in unpolar hydrocarbons
FC(F)(F)C(=O)OC(=O)C(F)(F)F
210.03
210.03
99+%

Chemical Identifiers

407-25-0
210.03
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
9845
FC(F)(F)C(=O)OC(=O)C(F)(F)F
C4F6O3
MFCD00000416
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu

GHS Signal Word: Danger

EINECSNumber : 206-982-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only