Learn More
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C
Supplier: Thermo Scientific Chemicals 169890011
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trifluoromethanesulfonic acid | |
-40.0°C | |
1.6900 g/mL | |
Authentic | |
Glass bottle | |
1.3260 to 1.3300 | |
MFCD00007514 | |
03,IV,34 | |
1.69 | |
trifluoromethanesulphonic acid, trimsylate, methanesulfonic acid, trifluoro, triflic acid, trifluoromethylsulfonic acid, perfluoromethanesulfonic acid, tfoh, trifluoromethane sulfonic acid, tfmsa, hotf | |
ITMCEJHCFYSIIV-UHFFFAOYSA-N | |
barium(2+) ditrifluoromethanesulfonate | |
62406 | |
CHEBI:48511 | |
99% | |
Fuming Liquid |
1493-13-6 | |
Colorless to Yellow | |
162.0°C | |
99% | |
CHF3O3S | |
CF3SO3H | |
1 L | |
04,532; 05,701; 06,617; 08,504; 09,531; 14,152; 15,41; 16,357; 17,24 | |
15,9842 | |
Solubility aq. soln.: soluble. Other solubilities: soluble in many polar organic solvents such as dim,sulfolane,dimethyl sulfoxide,dimethyl sulfone an | |
OS(=O)(=O)C(F)(F)F | |
150.07 | |
2.87 mPa.s (25°C) | |
150.07 | |
Extra Pure |
Chemical Identifiers
1493-13-6 | |
150.07 | |
ITMCEJHCFYSIIV-UHFFFAOYSA-N | |
62406 | |
OS(=O)(=O)C(F)(F)F |
CHF3O3S | |
MFCD00007514 | |
trifluoromethanesulphonic acid, trimsylate, methanesulfonic acid, trifluoro, triflic acid, trifluoromethylsulfonic acid, perfluoromethanesulfonic acid, tfoh, trifluoromethane sulfonic acid, tfmsa, hotf | |
CHEBI:48511 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
May be corrosive to metals.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 216-087-5
Recommended Storage : May slightly darken during storage
RUO – Research Use Only