missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic acid, 99%, extra pure

Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C | CAS: 1493-13-6 | CHF3O3S | 150.07 g/mol

$119.55 - $3691.46

Chemical Identifiers

CAS 1493-13-6
Molecular Formula CHF3O3S
Molecular Weight (g/mol) 150.07
MDL Number MFCD00007514
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
PubChem CID 62406
ChEBI CHEBI:48511
SMILES OS(=O)(=O)C(F)(F)F
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169890010
SDS View Documents
Thermo Scientific Chemicals
169890010
1 mL Glass bottle
Each for $119.55
Only null left
 
AC169890100
SDS View Documents
Thermo Scientific Chemicals
169890100
10 mL Glass bottle
Each for $123.60
Only null left
 
AC169890500
SDS View Documents
Thermo Scientific Chemicals
169890500
50 mL Glass bottle
Each for $383.63
Only null left
 
AC169892500
SDS View Documents
Thermo Scientific Chemicals
169892500
250 mL Glass bottle
Each for $1,298.93
Only null left
 
AC169890011
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
169890011
1 L Glass bottle
Each for $3,691.46
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1493-13-6
150.07
ITMCEJHCFYSIIV-UHFFFAOYSA-N
62406
OS(=O)(=O)C(F)(F)F
CHF3O3S
MFCD00007514
triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
CHEBI:48511

Specifications

1493-13-6
Colorless to Yellow
162.0°C
99%
CHF3O3S
CF3SO3H
1 mL
04,532; 05,701; 06,617; 08,504; 09,531; 14,152; 15,41; 16,357; 17,24
15,9842
Solubility in water: soluble. Other solubilities: soluble in many polar organic solvents such as dim,sulfolane,dimethyl sulfoxide,dimethyl sulfone an
OS(=O)(=O)C(F)(F)F
150.07
2.87 mPa.s (25°C)
150.07
Extra Pure
Trifluoromethanesulfonic acid
-40.0°C
1.6900g/mL
Authentic
Glass bottle
1.3260 to 1.3300
MFCD00007514
03,IV,34
1.69
triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
ITMCEJHCFYSIIV-UHFFFAOYSA-N
trifluoromethanesulfonic acid
62406
CHEBI:48511
99%
Fuming Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 216-087-5

Recommended Storage : May slightly darken during storage

SDS
Documents

Documents

RUO – Research Use Only