missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals

Alfa Aesar Trifluoromethanesulfonic anhydride, 98%, Quantity: 250g, Boiling Point: 81 deg.C to 83 deg.C, Melting Point: -80 deg.C, Molecular Weight: 282.127, Percent Purity: 98%, Beilstein: 1813600, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.72

$127.84 - $1285.63

Chemical Identifiers

CAS 358-23-6
Molecular Formula C2F6O5S2
Molecular Weight (g/mol) 282.127
MDL Number MFCD00000408
InChI Key WJKHJLXJJJATHN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
PubChem CID 67749
ChEBI CHEBI:48509
IUPAC Name trifluoromethylsulfonyl trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1176709
View Documents
Thermo Scientific Chemicals
A1176709
10 g
Each for $127.84
 
AAA1176714
View Documents
Thermo Scientific Chemicals
A1176714
25 g
Each for $252.01
 
AAA1176718
View Documents
Thermo Scientific Chemicals
A1176718
50 g
Each for $460.19
 
AAA1176722
View Documents
Thermo Scientific Chemicals
A1176722
100 g
Each for $836.38
 
AAA1176730
View Documents
Thermo Scientific Chemicals
A1176730
250 g
Each for $1,285.63
 
Description

Description

Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.

Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
Specifications

Chemical Identifiers

358-23-6
282.127
WJKHJLXJJJATHN-UHFFFAOYSA-N
67749
trifluoromethylsulfonyl trifluoromethanesulfonate
C2F6O5S2
MFCD00000408
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
CHEBI:48509
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F

Specifications

358-23-6
1.72
C2F6O5S2
(CF3SO2)2O
10 g
1813600
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
WJKHJLXJJJATHN-UHFFFAOYSA-N
trifluoromethylsulfonyl trifluoromethanesulfonate
67749
282.13
Trifluoromethanesulfonic anhydride
-80°C
81°C to 83°C
1.321
MFCD00000408
UN3265
Moisture sensitive
Miscible with dichloromethane. Immiscible with hydrocarbons.
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F
282.127
CHEBI:48509
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 206-616-8

RTECSNumber : PB2772000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only