missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triisopropyl phosphite, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A15247AK

 View more versions of this product

Catalog No. AAA15247AK


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Triisopropyl phosphite
0.908
46°C (114°F)
C9H21O3P
MFCD00008874
UN1993
Air and moisture sensitive
SJHCUXCOGGKFAI-UHFFFAOYSA-N
tripropan-2-yl phosphite
8304
≥90%
116-17-6
63°C to 64°C (11 mmHg)
Characteristic
1.411
250 mL
1701528
triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite
CC(C)OP(OC(C)C)OC(C)C
208.238
208.24

Chemical Identifiers

116-17-6
208.238
SJHCUXCOGGKFAI-UHFFFAOYSA-N
8304
CC(C)OP(OC(C)C)OC(C)C
C9H21O3P
MFCD00008874
triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite
tripropan-2-yl phosphite
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H341-H315-H319-H335
Flammable liquid and vapour.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c

H227-H301-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 204-130-0

RTECSNumber : TH2800000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only