Learn More
Triisopropyl phosphite, 90+%, Thermo Scientific Chemicals
$188.10 - $315.93
Chemical Identifiers
CAS | 116-17-6 |
---|---|
Molecular Formula | C9H21O3P |
Molecular Weight (g/mol) | 208.238 |
MDL Number | MFCD00008874 |
InChI Key | SJHCUXCOGGKFAI-UHFFFAOYSA-N |
Synonym | triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite |
PubChem CID | 8304 |
IUPAC Name | tripropan-2-yl phosphite |
SMILES | CC(C)OP(OC(C)C)OC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA15247AK
|
Thermo Scientific Chemicals
A15247AK |
250 mL |
Each for $188.10
|
|
|||||
AAA15247AP
|
Thermo Scientific Chemicals
A15247AP |
500 mL |
Each for $315.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
116-17-6 | |
208.238 | |
SJHCUXCOGGKFAI-UHFFFAOYSA-N | |
8304 | |
CC(C)OP(OC(C)C)OC(C)C |
C9H21O3P | |
MFCD00008874 | |
triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite | |
tripropan-2-yl phosphite |
Specifications
116-17-6 | |
63°C to 64°C (11 mmHg) | |
Characteristic | |
1.411 | |
250 mL | |
1701528 | |
triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite | |
CC(C)OP(OC(C)C)OC(C)C | |
208.238 | |
208.24 | |
Triisopropyl phosphite |
0.908 | |
46°C (114°F) | |
C9H21O3P | |
MFCD00008874 | |
UN1993 | |
Air and moisture sensitive | |
SJHCUXCOGGKFAI-UHFFFAOYSA-N | |
tripropan-2-yl phosphite | |
8304 | |
≥90% |
Safety and Handling
GHS H Statement
H226-H341-H315-H319-H335
Flammable liquid and vapour.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
H227-H301-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 204-130-0
RTECSNumber : TH2800000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only