missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethoxy(4-vinylphenyl)silane 97.0+%, TCI America™
$242.27 - $726.83
Chemical Identifiers
CAS | 18001-13-3 |
---|---|
Molecular Formula | C11H16O3Si |
Molecular Weight (g/mol) | 224.33 |
MDL Number | MFCD21596662 |
InChI Key | LTQBNYCMVZQRSD-UHFFFAOYSA-N |
PubChem CID | 15606295 |
IUPAC Name | (4-ethenylphenyl)trimethoxysilane |
SMILES | CO[Si](OC)(OC)C1=CC=C(C=C)C=C1 |
Chemical Identifiers
18001-13-3 | |
224.33 | |
LTQBNYCMVZQRSD-UHFFFAOYSA-N | |
(4-ethenylphenyl)trimethoxysilane |
C11H16O3Si | |
MFCD21596662 | |
15606295 | |
CO[Si](OC)(OC)C1=CC=C(C=C)C=C1 |
Specifications
18001-13-3 | |
77°C | |
MFCD21596662 | |
LTQBNYCMVZQRSD-UHFFFAOYSA-N | |
(4-ethenylphenyl)trimethoxysilane | |
15606295 | |
≥97.0% (GC) | |
Trimethoxy(4-vinylphenyl)silane |
Yellow | |
C11H16O3Si | |
5 g | |
CO[Si](OC)(OC)C1=CC=C(C=C)C=C1 | |
224.33 | |
224.33 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-4371
TSCA : No