Learn More
Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals
$166.07 - $591.48
Chemical Identifiers
CAS | 75-50-3 |
---|---|
Molecular Formula | C3H9N |
Molecular Weight (g/mol) | 59.11 |
MDL Number | MFCD00008327 |
InChI Key | GETQZCLCWQTVFV-UHFFFAOYSA-N |
Synonym | trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 |
PubChem CID | 1146 |
ChEBI | CHEBI:18139 |
SMILES | CN(C)C |
Description
Trimethylamine solution used in research. As flavoring agents and adjuvants. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators, strongly basic anion exchange resins, dye leveling agents and a number of basic dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-50-3 | |
59.11 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
1146 | |
CN(C)C |
C3H9N | |
MFCD00008327 | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
CHEBI:18139 |
Specifications
75-50-3 | |
Colorless | |
−20°C (−4°F) | |
C3H9N | |
500 mL | |
956566 | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
trimethylamine | |
1146 | |
59.11 | |
Liquid |
-2°C | |
30°C | |
Unpleasant | |
MFCD00008327 | |
UN1297 | |
14,9710 | |
Soluble In Water,8.9e+005mg/L. | |
CN(C)C | |
59.11 | |
CHEBI:18139 | |
45% w/w aq. soln. | |
Trimethylamine |
Safety and Handling
GHS H Statement
H224-H314-H302-H332-H335
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H224-H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: TRIMETHYLAMINE, AQUEOUS SOLUTIONS
EINECSNumber : 200-875-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only