missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2685714

 View more versions of this product

Catalog No. AAH2685714


Explore available promotions

Description

Description

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
1.27
26°C (79°F)
1.367
25 g
trimethylsilyl 2,2-difluoro-2-fluorosulfonyl acetate, trimethylsilyl 2-fluorosulfonyl difluoroacetate, trimethylsilyl2-fluorosulphonyl difluoroacetate, trimethylsilyl fluorosulfonyldifluoroacetate, trimethylsilyl difluoro fluorosulfonyl acetate, trimethylsilyl 2-fluorosulphonyl difluoroacetate, 2,2-difluoro-2-fluorosulfonyl acetic acid trimethylsilyl ester, pubchem22642, g00021-watson-int, acmc-1c7yr
XHVSCKNABCCCAC-UHFFFAOYSA-N
trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate
2778022
94%
120801-75-4
156°C
C5H9F3O4SSi
MFCD02093343
UN2920
Reacts with water.
C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O
250.26
250.27

Chemical Identifiers

120801-75-4
250.26
XHVSCKNABCCCAC-UHFFFAOYSA-N
trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate
C5H9F3O4SSi
MFCD02093343
2778022
C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only