Learn More
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%, Thermo Scientific Chemicals
$130.59 - $433.84
Chemical Identifiers
CAS | 120801-75-4 |
---|---|
Molecular Formula | C5H9F3O4SSi |
Molecular Weight (g/mol) | 250.26 |
MDL Number | MFCD02093343 |
InChI Key | XHVSCKNABCCCAC-UHFFFAOYSA-N |
PubChem CID | 2778022 |
IUPAC Name | trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate |
SMILES | C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2685706
|
Thermo Scientific Chemicals
H2685706 |
5 g |
Each for $130.59
|
|
|||||
AAH2685714
|
Thermo Scientific Chemicals
H2685714 |
25 g |
Each for $433.84
|
|
|||||
Description
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.
Solubility
Reacts with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
120801-75-4 | |
250.26 | |
XHVSCKNABCCCAC-UHFFFAOYSA-N | |
trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate |
C5H9F3O4SSi | |
MFCD02093343 | |
2778022 | |
C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O |
Specifications
120801-75-4 | |
156°C | |
C5H9F3O4SSi | |
MFCD02093343 | |
UN2920 | |
Reacts with water. | |
C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O | |
250.26 | |
250.27 | |
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate |
1.27 | |
26°C (79°F) | |
1.367 | |
5 g | |
trimethylsilyl 2,2-difluoro-2-fluorosulfonyl acetate, trimethylsilyl 2-fluorosulfonyl difluoroacetate, trimethylsilyl2-fluorosulphonyl difluoroacetate, trimethylsilyl fluorosulfonyldifluoroacetate, trimethylsilyl difluoro fluorosulfonyl acetate, trimethylsilyl 2-fluorosulphonyl difluoroacetate, 2,2-difluoro-2-fluorosulfonyl acetic acid trimethylsilyl ester, pubchem22642, g00021-watson-int, acmc-1c7yr | |
XHVSCKNABCCCAC-UHFFFAOYSA-N | |
trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate | |
2778022 | |
94% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only