missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsulfoxonium iodide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 140261000

 View more versions of this product

Catalog No. AC140261000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trimethylsulfoxonium iodide
1774-47-6
White to Yellow
98%
C3H9IOS
MFCD00011899
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1
BPLKQGGAXWRFOE-UHFFFAOYSA-M
trimethyl(oxo)-λ⁶-sulfanylium iodide
74498
98%
98%
175.0°C
Authentic
Plastic bottle
(CH3)3S(O)I
100 g
Solubility in water: 15g/L (20°C). Other solubilities: insoluble in ethanol and ether
[I-].C[S+](C)(C)=O
220.07
220.07
Crystalline Powder

Chemical Identifiers

1774-47-6
220.07
BPLKQGGAXWRFOE-UHFFFAOYSA-M
74498
[I-].C[S+](C)(C)=O
C3H9IOS
MFCD00011899
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1
trimethyl(oxo)-λ⁶-sulfanylium iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 217-204-2

RTECSNumber : WS3585000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only