missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsulfoxonium iodide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1458922

 View more versions of this product

Catalog No. AAA1458922


Explore available promotions

Description

Description

Trimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases. Light sensitive.
Specifications

Specifications

Trimethylsulfoxonium iodide
∼170°C (decomposition)
MFCD00011899
3595854
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1
BPLKQGGAXWRFOE-UHFFFAOYSA-M
trimethyl(oxo)-λ⁶-sulfanylium iodide
74498
≥98%
1774-47-6
C3H9IOS
100 g
Light sensitive
Soluble in water.
[I-].C[S+](C)(C)=O
220.07
220.07

Chemical Identifiers

1774-47-6
220.07
BPLKQGGAXWRFOE-UHFFFAOYSA-M
74498
[I-].C[S+](C)(C)=O
C3H9IOS
MFCD00011899
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1
trimethyl(oxo)-λ⁶-sulfanylium iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 217-204-2

RTECSNumber : WS3585000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only