Learn More
Trimethylsulfoxonium iodide, 98+%, Thermo Scientific Chemicals
$44.58 - $328.71
Chemical Identifiers
CAS | 1774-47-6 |
---|---|
Molecular Formula | C3H9IOS |
Molecular Weight (g/mol) | 220.07 |
MDL Number | MFCD00011899 |
InChI Key | BPLKQGGAXWRFOE-UHFFFAOYSA-M |
Synonym | trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1 |
PubChem CID | 74498 |
IUPAC Name | trimethyl(oxo)-λâ¶-sulfanylium iodide |
SMILES | [I-].C[S+](C)(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1458914
|
Thermo Scientific Chemicals
A1458914 |
25 g |
Each for $44.58
|
|
|||||
AAA1458922
|
Thermo Scientific Chemicals
A1458922 |
100 g |
Each for $100.99
|
|
|||||
AAA1458936
|
Thermo Scientific Chemicals
A1458936 |
500 g |
Each for $328.71
|
|
|||||
Description
Trimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Light sensitive.
Chemical Identifiers
1774-47-6 | |
220.07 | |
BPLKQGGAXWRFOE-UHFFFAOYSA-M | |
74498 | |
[I-].C[S+](C)(C)=O |
C3H9IOS | |
MFCD00011899 | |
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1 | |
trimethyl(oxo)-λâ¶-sulfanylium iodide |
Specifications
1774-47-6 | |
C3H9IOS | |
25 g | |
Light sensitive | |
Soluble in water. | |
[I-].C[S+](C)(C)=O | |
220.07 | |
220.07 | |
Trimethylsulfoxonium iodide |
∼170°C (decomposition) | |
MFCD00011899 | |
3595854 | |
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1 | |
BPLKQGGAXWRFOE-UHFFFAOYSA-M | |
trimethyl(oxo)-λâ¶-sulfanylium iodide | |
74498 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 217-204-2
RTECSNumber : WS3585000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only