missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triphenylmethanol 99.0+%, TCI America™
Supplier: TCI America T051625G
Specifications
Triphenylmethanol | |
163°C | |
C19H16O | |
25 g | |
LZTRCELOJRDYMQ-UHFFFAOYSA-N | |
triphenylmethanol | |
6457 | |
≥99.0% (GC) |
76-84-6 | |
380°C | |
MFCD00004445,MFCD10565638 | |
triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl | |
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
260.34 | |
260.34 | |
Crystalline Powder |
Chemical Identifiers
76-84-6 | |
260.34 | |
LZTRCELOJRDYMQ-UHFFFAOYSA-N | |
6457 | |
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C19H16O | |
MFCD00004445,MFCD10565638 | |
triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl | |
triphenylmethanol |
Safety and Handling
TSCA : Yes