Learn More
Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 291070050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Triphenylphosphine dibromide | |
1034-39-5 | |
Pink to Yellow | |
Plastic bottle | |
(C6H5)3PBr2 | |
5 g | |
01,1247; 13,333 | |
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane | |
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
422.10 | |
422.09 | |
Crystalline Powder |
ca. 33% bromine | |
235.0°C | |
Authentic | |
C18H15Br2P | |
MFCD00000054 | |
16, 864 | |
15, 9917 | |
OCXGTPDKNBIOTF-UHFFFAOYSA-N | |
dibromo(triphenyl)-$l^{5}-phosphane | |
70579 | |
ca. 33% bromine |
Chemical Identifiers
1034-39-5 | |
422.10 | |
OCXGTPDKNBIOTF-UHFFFAOYSA-N | |
70579 | |
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C18H15Br2P | |
MFCD00000054 | |
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane | |
dibromo(triphenyl)-$l^{5}-phosphane |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 213-855-1
RUO – Research Use Only