missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals

$64.06 - $191.79

Chemical Identifiers

CAS 1034-39-5
Molecular Formula C18H15Br2P
Molecular Weight (g/mol) 422.10
MDL Number MFCD00000054
InChI Key OCXGTPDKNBIOTF-UHFFFAOYSA-N
Synonym dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane
PubChem CID 70579
IUPAC Name dibromo(triphenyl)-$l^{5}-phosphane
SMILES BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC291070050
SDS View Documents
Thermo Scientific Chemicals
291070050
5 g Plastic bottle
Each for $64.06
 
AC291070250
SDS View Documents
Thermo Scientific Chemicals
291070250
25 g Plastic bottle
Each for $191.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

1034-39-5
422.10
OCXGTPDKNBIOTF-UHFFFAOYSA-N
70579
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C18H15Br2P
MFCD00000054
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane
dibromo(triphenyl)-$l^{5}-phosphane

Specifications

1034-39-5
Pink to Yellow
Plastic bottle
(C6H5)3PBr2
5 g
01,1247; 13,333
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
422.10
422.09
Crystalline Powder
235.0°C
Authentic
C18H15Br2P
MFCD00000054
16, 864
15, 9917
OCXGTPDKNBIOTF-UHFFFAOYSA-N
dibromo(triphenyl)-$l^{5}-phosphane
70579
ca. 33% bromine
Triphenylphosphine dibromide, ca. 33% bromine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-855-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only