missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphosgene 98.0+%, TCI America™

Supplier:  TCI America T146725G

 View more versions of this product

Catalog No. T146725G


Explore available promotions

Description

Description

Specifications

Specifications

Triphosgene
80°C
206°C
MFCD00062848
2928
UCPYLLCMEDAXFR-UHFFFAOYSA-N
bis(trichloromethyl) carbonate
94429
≥98.0% (T)
32315-10-9
White-Yellow
C3Cl6O3
25 g
triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl
296.73
296.73
Crystalline Powder

Chemical Identifiers

32315-10-9
296.73
UCPYLLCMEDAXFR-UHFFFAOYSA-N
94429
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl
C3Cl6O3
MFCD00062848
triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene
bis(trichloromethyl) carbonate
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions