missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tripropionin, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 275102500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tripropionin | |
139-45-7 | |
1.0820g/mL | |
162°C | |
96% min. (GC) | |
C12H20O6 | |
MFCD00027005 | |
1.082 | |
YZWRNSARCRTXDS-UHFFFAOYSA-N | |
2,3-di(propanoyloxy)propyl propanoate | |
8763 | |
97% |
97% | |
Colorless | |
151°C (2.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4310 to 1.434 | |
250 g | |
tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride | |
CCC(=O)OCC(COC(=O)CC)OC(=O)CC | |
260.28 | |
260.28 | |
Liquid |
Chemical Identifiers
139-45-7 | |
260.28 | |
YZWRNSARCRTXDS-UHFFFAOYSA-N | |
8763 | |
CCC(=O)OCC(COC(=O)CC)OC(=O)CC |
C12H20O6 | |
MFCD00027005 | |
tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride | |
2,3-di(propanoyloxy)propyl propanoate |
Safety and Handling
LIGHT SENSITIVE
EINECSNumber : 205-365-1
RTECSNumber : YK6000000
TSCA : TSCA
RUO – Research Use Only